Learn More
Sigma Aldrich Fine Chemicals Biosciences KontesR Williamson microsc
Catalog No. 502900428
Encompass
Kit contains glassware needed to perform experiments described in the textbook Macroscale and Microscale Organic Experiments. This economical microscale glassware kit has a unique positive connection without the use of threads or ST joints. Glassware is supplied in a foam-lined corrugated cardboard Ekono-Case.
Catalog No. 50-290-0428 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2566331KT
Kit contains glassware needed to perform experiments described in the textbook Macroscale and Microscale Organic Experiments. This economical microscale glassware kit has a unique positive connection without the use of threads or ST joints. Glassware is supplied in a foam-lined corrugated cardboard Ekono-Case.