Learn More
Cellucap K100 18 in. Koronet Hairnets
Catalog No. 19133811 Shop All Cellucap Products
This item is not returnable. View return policy
Catalog No. 19-133-811 Supplier Cellucap Supplier No. K10018WH
Add to Cart
Add to Cart
This item is not returnable. View return policy
Honeycomb hairnets are a standard in food processing, safety, and industrial applications.
  • 18 in. diameter
  • Available in white or brown
  • Single strand 30 denier net is made with a 0.125 in. aperture for better hair containment
  • Domestic boxed
Specifications
Product Type Hairnet
Material Polyester
Disposable Single-use
Model K10018
Packaging Type Boxed