Learn More
TARGETMOL CHEMICALS INC KOUMINE 20MG
SDP
Catalog No. 502579719
Encompass_Preferred
Also available in 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Koumine can treat the diabetic neuropathy. 2. Koumine has antineoplastic effect may be a future breast cancer chemotherapeutic agent. 3. Koumine has a significant analgesic effect in rodent behavioral models of inflammatory and neuropathic pain the mechanism may be associated with the upregulation of allopregnanolone in the spinal cord. purity: 99%
Catalog No. 50-257-9719 Supplier TARGETMOL CHEMICALS INC Supplier No. T5S066120MG
Also available in 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Koumine can treat the diabetic neuropathy. 2. Koumine has antineoplastic effect may be a future breast cancer chemotherapeutic agent. 3. Koumine has a significant analgesic effect in rodent behavioral models of inflammatory and neuropathic pain the mechanism may be associated with the upregulation of allopregnanolone in the spinal cord. purity: 99%