Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kova Plastics by Alltrista KO-LEC-PAC™ Urine Specimen Collection and Transport
GSA_VA

Catalog No. 22749741
Change view
Click to view available options
No. per Pack:
500/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
22-749741 500/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-749741 Supplier Kova Plastics by Alltrista Supplier No. 87141E
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Complete with specially engineered microscopic slides, sample tubes, labels, collection cups and transport racks for reliable, simple and effective workflow standardization

  • Complete kit for collection and secure transportation of up to 500 urine samples

Package contains:

  • Cups: Used for collection of a 12mL specimen
  • Super Tubes: A 12mL graduated non-sterile disposable centrifuge tube manufactured of high-quality, break-resistant clear plastic and marked in 1mL increments
  • Caps: Provides secure closure to prevent spillage during transportation to the lab, as well as aerosol contamination during centrifugation
  • Labels
  • Transport Racks: Used to keep tubes secure and upright during transportation

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Product Type Urine Cytology Collection and Transport Kit
For Use With (Application) Collection and secure transportation of urine samples
Includes 500 Super Tubes, 500 Caps, 500 cups, 500 labels and five transport racks
No. per Pack 500/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.