Learn More
Kova Plastics by Alltrista System Super Pac 1000 with Caps
Catalog No. 22286140
Change view
Click to view available options
No. per Pack:
1000 Pk.
Catalog No. No. per Pack
22-286140 1000 Pk.
1 options
Catalog No. 22-286140 Supplier Kova Plastics by Alltrista Supplier No. 87153E
Add to Cart
Add to Cart
Complete with specially engineered microscopic slides (non-gridded), sample tubes and caps, and custom pipettes or petters for reliable, simple and effective workflow standardization
  • Complete urinalysis kit for up to 1000 determination
Package contains:
  • Super Tubes(1000): 12mL graduated non-sterile disposable centrifuge tube manufactured from high quality, break-resistant clear plastic and marked in 1mL increments
  • Caps (1000): Provides secure closures during transport and storage, as well as reduces the risk of aerosoling of potentially infectious specimens during centrifugation
  • Petters (1000): A disposable plastic transfer pipette that features a unique lock for one-step, contamination-free decanting, retaining 1.0mL of urine after centrifugation
  • Glasstic™ Slides 10 (100): A precision manufactured, 10-chambered disposable microscope slide made of optically clear plastic (non-gridded).
Specifications
Product Type Urine Collection Tube
Includes 100 Glasstic™ 10-chambered slides (non-gridded),1000 Petters, 1000 Super Tubes and caps
No. per Pack 1000 Pk.