Learn More
Kova Plastics by Alltrista System Value Pac 500
GSA_VA
Catalog No. 22749743
Change view
Click to view available options
Volume (Metric):
500 Determinations
Catalog No. Volume (Metric)
22-749743 500 Determinations
1 options
This item is not returnable. View return policy
Catalog No. 22-749743 Supplier Kova Plastics by Alltrista Supplier No. 87156E
Add to Cart
Add to Cart
This item is not returnable. View return policy
Complete with specially engineered microscopic slides, sample tubes, custom pipettes or petters, and collection cups for reliable, simple and effective workflow standardization
Complete with specially engineered microscopic slides, sample tubes, custom pipettes or petters , and collection cups for reliable, simple and effective workflow standardization
  • Complete urinalysis kit for up to 500 determinations
Package contains:
  • Glasstic™ Slides 10 (50): A precision manufactured, 10-chambered disposable microscope slide made of optically clear plastic, with hemocytometer counting grid. The Glasstic slide grid is in the focal plane of the specimen, allowing for use over a range of magnifications
  • Petters (500): A disposable plastic transfer pipette that features a unique lock for one-step, contamination-free decanting, retaining 1.0mL of urine after centrifugation
  • Economy Tubes (500): Tubes for urine collection and analysis made from break-resistant polystyrene
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Product Type Urine Collection Kit
Disposable Single-use
Material Plastic
Includes 50 Glasstic 10-chambered slides, 500 Petters and 500 economy tubes standardization
No. per Pack 500 Pk.
Volume (Metric) 500 Determinations