Learn More
Kova Plastics by Alltrista System Super Pac 1000
Catalog No. 22749820
Change view
Click to view available options
No. per Pack:
1000 Pk.
Catalog No. No. per Pack
22-749820 1000 Pk.
1 options
This item is not returnable. View return policy
Catalog No. 22-749820 Supplier Kova Plastics by Alltrista Supplier No. 87154E
Add to Cart
Add to Cart
This item is not returnable. View return policy
Complete with specially engineered microscopic slides and sample tube for reliable, simple and effective workflow standardization
  • Complete urinalysis kit for up to 1000 determinations
Package contains:
  • Super Tubes (1000): A 12mL graduated non-sterile disposable centrifuge tube manufactured of high-quality, break-resistant clear plastic and marked in 1mL increments
  • Petters (1000): A disposable plastic transfer pipette that features a unique lock for one-step, contamination-free decanting, retaining 1.0mL of urine after centrifugation
  • Glasstic™ Slides 10 (100): A precision manufactured, 10-chambered disposable microscope slide made of optically clear plastic. The Glasstic slide grid is in the focal plane of the specimen, allowing for use over a range of magnifications
Specifications
Product Type Urine Collection Tube
Includes 100 Glasstic 10-chambered slides,1000 Petters, 1000 Super Tubes (without caps)
No. per Pack 1000 Pk.