Learn More
Kova Plastics by Alltrista Petters
GSA_VA
Catalog No. VT87135
Change view
Click to view available options
Disposable:
Single-use
Catalog No. Disposable
VT87135 Single-use
1 options
This item is not returnable. View return policy
Catalog No. VT87135 Supplier Kova Plastics by Alltrista Supplier No. 87135E
Add to Cart
Add to Cart
This item is not returnable. View return policy
Multipurpose plastic transfer pipette designed for contamination-free decanting
  • Proprietary design
  • Assures volume consistent sediment collection and suspension with one-step, contamination-free decanting
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Product Type Urine Collection Tube
Disposable Single-use
Material Plastic
For Use With (Application) Urine Collection and Analysis