Learn More
Kova Plastics by Alltrista Tube Caps
GSA_VA
Catalog No. VT87139
Change view
Click to view available options
No. per Pack:
500/Pk.
Catalog No. No. per Pack
VT87139 500/Pk.
1 options
This item is not returnable. View return policy
Catalog No. VT87139 Supplier Kova Plastics by Alltrista Supplier No. 87139E
Add to Cart
Add to Cart
This item is not returnable. View return policy
Tube Caps provide secure fit during transport and storage of specimens
  • Fits both Super and Economy Tubes
  • Prevents spillage during specimen collection, transportation, and handling, minimizing biohazardous exposure
  • Prevents aerosoling of potentially infectious specimens during centrifugation
  • Convenient flip cap cover one-third of cap circumference
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Product Type Urine Collection Tube Cap
Closure Type Flex with Flip Tab
For Use With (Application) Urine Collection and Analysis
No. per Pack 500/Pk.