Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kovac's Aldehyde Reagent, for Detection of Indole Producing Bacteria, Ricca Chemical
SDP

Catalog No. 426016 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4260-16 500 mL
4260-4 120 mL
4260-1 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 4260-16 Supplier Ricca Chemical Company Supplier No. 426016
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Detection of Indole Producing Bacteria

Specifications

Chemical Name or Material Kovac's Aldehyde Reagent
Color Green-Yellow
CAS 71-36-3, 7647-01-0, 100-10-7
CAS Min % 63.0, 30.35, 5.15
CAS Max % 65.0, 31.59, 5.36
Grade Laboratory
Packaging Amber Glass Bottle
Quantity 500 mL
Physical Form Heavy Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.