Learn More
Trajan Scientific Clear QsertVial With Fused 300l insert,12x32mm Screw Kit Pack
Catalog No. 50635224
Encompass_Preferred
Kit Pack. QsertVial, 12x32mm, Screw, Clear With fused 300µl insert, pre-slit, PTFE/Silicone, Blue QuikCap, 100 Pack
Catalog No. 50-635-224 Supplier Trajan Scientific Supplier No. XQS2042
Kit Pack. QsertVial, 12x32mm, Screw, Clear With fused 300µl insert, pre-slit, PTFE/Silicone, Blue QuikCap, 100 Pack