Learn More
Trajan Scientific Kit Pack Screw PP 300l, 12x32mm, Pre-slit PTFE/Silicone, 100 Pack
Catalog No. 50635206
Encompass_Preferred
Kit Pack, Screw, PP, 12x32mm, Flat bottom, Quantity100 Pack No, QuikCap/QuiKVial
Catalog No. 50-635-206 Supplier Trajan Scientific Supplier No. XAR6035
Kit Pack, Screw, PP, 12x32mm, Flat bottom, Quantity100 Pack No, QuikCap/QuiKVial