Learn More
Selleck Chemical LLC KP-457-E1386-5MG
SDP
Catalog No. 502389848
Encompass_Preferred
KP-457 (TNF- -converting enzyme) is a selective inhibitor of ADAM17 with IC50 of 11 1 nM/L (cell-free assay) It also cleaves various molecules such as GPIb GPV and TNF-
Catalog No. 50-238-9848 Supplier Selleck Chemical LLC Supplier No. E13865MG
Add to Cart
Add to Cart
KP-457 (TNF- -converting enzyme) is a selective inhibitor of ADAM17 with IC50 of 11 1 nM/L (cell-free assay) It also cleaves various molecules such as GPIb GPV and TNF-