Learn More
Medchemexpress LLC KP-457 (GMP) | 1365803-52-6 | 480.55 | 100 MG
SDP
Catalog No. 5000343210
Encompass_Preferred
KP-457 (GMP) is a selective a disintegrin and metalloproteinase 17 (ADAM17) inhibitor. Produced using GMP guidelines, this small molecule serves as an auxiliary reagent for cell therapy manufacture, offering higher selectivity for ADAM17 compared to other MMPs and ADAM10.
  • Selective ADAM17 inhibitor
  • Manufactured under GMP guidelines
  • Auxiliary reagent for cell therapy
  • Demonstrates higher selectivity for ADAM17 over other MMPs and ADAM10
Catalog No. 50-003-43210 Supplier Medchemexpress LLC Supplier No. HY110397G10MG
KP-457 (GMP) is a selective a disintegrin and metalloproteinase 17 (ADAM17) inhibitor. Produced using GMP guidelines, this small molecule serves as an auxiliary reagent for cell therapy manufacture, offering higher selectivity for ADAM17 compared to other MMPs and ADAM10.
  • Selective ADAM17 inhibitor
  • Manufactured under GMP guidelines
  • Auxiliary reagent for cell therapy
  • Demonstrates higher selectivity for ADAM17 over other MMPs and ADAM10