Learn More
MERCEDES SCIENTIFIC KP Marker 12/bx - The KP Marker comes in a BX/12. This laboratory marking pen is excellent for writing on cassettes and glass as the KP Marker is resistant to solvents.
Catalog No. NC3450222
Encompass
KP Marker 12/bx - The KP Marker comes in a BX/12. This laboratory marking pen is excellent for writing on cassettes and glass as the KP Marker is resistant to solvents.
This item is not returnable. View return policy
Catalog No. NC3450222 Supplier MERCEDES SCIENTIFIC Supplier No. KLI KPMARKER
KP Marker 12/bx - The KP Marker comes in a BX/12. This laboratory marking pen is excellent for writing on cassettes and glass as the KP Marker is resistant to solvents.