Learn More
Biotage TLC Plates, 5cm x 10cm Glass, with Amino silica, 50 per cas
SDP
Catalog No. NC0891484
Encompass
Biotage TLC plates are the perfect tool to predict flash methods for Biotage Sfar columns. For synthetic chemists, the benefits of matched TLC plates and flash cartridges are better purification throughput, increased compound purity and yield, and reduced solvent cost. Use these plates with Biotage Sfar KP-Amino columns.These glass plates are coated with amino functionalize silica and are 5cm x 10cm packed 50 plates to a case.
Catalog No. NC0891484 Supplier Biotage Supplier No. TLCKPNH0510FI
Biotage TLC plates are the perfect tool to predict flash methods for Biotage Sfar columns. For synthetic chemists, the benefits of matched TLC plates and flash cartridges are better purification throughput, increased compound purity and yield, and reduced solvent cost. Use these plates with Biotage Sfar KP-Amino columns.These glass plates are coated with amino functionalize silica and are 5cm x 10cm packed 50 plates to a case.