Learn More
Trajan Scientific Amber QsertVial Screw With Fused 300l insert, Kit Pack
Catalog No. 50635227
Encompass_Preferred
Kit Pack. QsertVial, 12x32mm, Screw, Amber With fused 300µl insert, pre-slit, PTFE/Silicone, Blue QuikCap, 100 Pack
Catalog No. 50-635-227 Supplier Trajan Scientific Supplier No. XQS2044
Kit Pack. QsertVial, 12x32mm, Screw, Amber With fused 300µl insert, pre-slit, PTFE/Silicone, Blue QuikCap, 100 Pack