Learn More
Creative Bioarray KP4,PANCREATIC DUCTAL CELL
Catalog No. NC2221702
Encompass
Pancreatic ductal cell carcinoma. Mother cell line of KP4-1, -2, and -3
This item is not returnable. View return policy
Catalog No. NC2221702 Supplier Creative Bioarray Supplier No. CSCC6433J
Pancreatic ductal cell carcinoma. Mother cell line of KP4-1, -2, and -3