Learn More
Key Scientific Products KPC,MLB AND OXA-48 CONFIRM Kit, 50 per Pack
SDP
Catalog No. NC1047666
Encompass_Preferred
This kit is a multi disc method that allows the identification and differentiation of KPC and MBL positive organisms. Carbapenem resistance may also be due to impermeability or the presence of the enzyme OXA-48. The OXA-48 enzyme can be confirmed through the addition of a Temocillin 30ug disc to the ID Kit. This ID Kit contains four discs which all contain 10ug meropenem. In addition, 3 of the discs also contain inhibitors of various B-lactamases. By comparing the various inhibition zone sizes produced by the suspected organism it is possible to identify the presence of AmpC, MBL and KPC enzymes. As all OXA-48 producing Enterobacteriaceae are highly resistant to Temocillin they will exhibit no zone of inhibition around the Temocillin disc.
Catalog No. NC1047666 Supplier Key Scientific Products Supplier No. RO98015
This kit is a multi disc method that allows the identification and differentiation of KPC and MBL positive organisms. Carbapenem resistance may also be due to impermeability or the presence of the enzyme OXA-48. The OXA-48 enzyme can be confirmed through the addition of a Temocillin 30ug disc to the ID Kit. This ID Kit contains four discs which all contain 10ug meropenem. In addition, 3 of the discs also contain inhibitors of various B-lactamases. By comparing the various inhibition zone sizes produced by the suspected organism it is possible to identify the presence of AmpC, MBL and KPC enzymes. As all OXA-48 producing Enterobacteriaceae are highly resistant to Temocillin they will exhibit no zone of inhibition around the Temocillin disc.