Learn More
Medchemexpress LLC HY-128578 5mg Medchemexpress, KPLH1130 CAS:906669-07-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1285785MG
Encompass_Preferred
Medchemexpress, HY-128578 5mg KPLH1130 CAS:906669-07-6 KPLH1130 is a specific pyruvate dehydrogenase kinase (PDK) inhibitor, blocks macrophage polarization and attenuates proinflammatory responses. KPLH1130 improves glucose tolerance in HFD-fed mice. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9753