Learn More
Medchemexpress LLC KPLH1130 | 906669-07-6 | 99.9% | 1 ML
SDP
Catalog No. 5000341960
Encompass_Preferred
KPLH1130 is a pyruvate dehydrogenase kinase (PDK) inhibitor. It inhibits M1 macrophage polarization by decreasing pro-inflammatory cytokines and reducing M1 phenotype markers. It prevents the decrease in mitochondrial oxygen consumption rate caused by inflammatory stimuli and improves glucose tolerance in mice fed a high-fat diet. This compound is useful for studying obesity-associated metabolic disorders and inflammatory conditions.
  • Inhibits M1 macrophage polarization
  • Decreases expression of pro-inflammatory cytokines
  • Reduces M1 phenotype markers (HIF-1α, iNOS)
  • Lowers nitric oxide (NO) production
  • Prevents decrease in mitochondrial oxygen consumption rate (OCR) induced by inflammatory stimuli
  • Improves glucose tolerance in high-fat diet-fed mice
  • Useful for studying obesity-associated metabolic disorders
  • Useful for studying inflammatory conditions
Catalog No. 50-003-41960 Supplier Medchemexpress LLC Supplier No. HY12857810MM/1ML
KPLH1130 is a pyruvate dehydrogenase kinase (PDK) inhibitor. It inhibits M1 macrophage polarization by decreasing pro-inflammatory cytokines and reducing M1 phenotype markers. It prevents the decrease in mitochondrial oxygen consumption rate caused by inflammatory stimuli and improves glucose tolerance in mice fed a high-fat diet. This compound is useful for studying obesity-associated metabolic disorders and inflammatory conditions.
  • Inhibits M1 macrophage polarization
  • Decreases expression of pro-inflammatory cytokines
  • Reduces M1 phenotype markers (HIF-1α, iNOS)
  • Lowers nitric oxide (NO) production
  • Prevents decrease in mitochondrial oxygen consumption rate (OCR) induced by inflammatory stimuli
  • Improves glucose tolerance in high-fat diet-fed mice
  • Useful for studying obesity-associated metabolic disorders
  • Useful for studying inflammatory conditions