Learn More
Medchemexpress LLC KPLH1130 | 906669-07-6 | 99.9% | 50 MG
SDP
Catalog No. 5000341258
Encompass_Preferred
KPLH1130 is a pyruvate dehydrogenase kinase (PDK) inhibitor. It potently inhibits M1 macrophage polarization by reducing pro-inflammatory cytokines, decreasing M1 phenotype markers (HIF-1α, iNOS), and lowering nitric oxide (NO) production. It prevents the reduction of mitochondrial oxygen consumption rate (OCR) induced by inflammatory stimuli and improves glucose tolerance in HFD-fed mice. It can be used for the study of obesity-associated metabolic disorders and other inflammatory conditions.
  • Potently inhibits M1 macrophage polarization
  • Reduces pro-inflammatory cytokines
  • Decreases M1 phenotype markers (HIF-1α, iNOS)
  • Lowers nitric oxide (NO) production
  • Prevents reduction of mitochondrial oxygen consumption rate (OCR)
  • Improves glucose tolerance in HFD-fed mice
  • Useful for studying obesity-associated metabolic disorders
  • Useful for studying inflammatory conditions
Catalog No. 50-003-41258 Supplier Medchemexpress LLC Supplier No. HY12857850MG
KPLH1130 is a pyruvate dehydrogenase kinase (PDK) inhibitor. It potently inhibits M1 macrophage polarization by reducing pro-inflammatory cytokines, decreasing M1 phenotype markers (HIF-1α, iNOS), and lowering nitric oxide (NO) production. It prevents the reduction of mitochondrial oxygen consumption rate (OCR) induced by inflammatory stimuli and improves glucose tolerance in HFD-fed mice. It can be used for the study of obesity-associated metabolic disorders and other inflammatory conditions.
  • Potently inhibits M1 macrophage polarization
  • Reduces pro-inflammatory cytokines
  • Decreases M1 phenotype markers (HIF-1α, iNOS)
  • Lowers nitric oxide (NO) production
  • Prevents reduction of mitochondrial oxygen consumption rate (OCR)
  • Improves glucose tolerance in HFD-fed mice
  • Useful for studying obesity-associated metabolic disorders
  • Useful for studying inflammatory conditions