Learn More
Aviva Systems Biology Corp KPNA1 Antibody - N-terminal region (ARP54670_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54670_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KPNA1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KPNA1. Alternate names: IPOA5, NPI-1, RCH2, SRP1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61277