Learn More
Aviva Systems Biology Corp KPNA2 Antibody - middle region (ARP54673_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54673_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KPNA2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KPNA2. Alternate names: IPOA1, QIP2, RCH1, SRP1alphaApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-61279