Learn More
Aviva Systems Biology Corp KPNA3 Antibody - N-terminal region (ARP54674_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54674_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KPNA3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KPNA3. Alternate names: IPOA4, SRP1gamma, SRP4, hSRP1, SRP1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-61280