Learn More
Aviva Systems Biology Corp KPNA4 Antibody - N-terminal region (ARP54676_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54676_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KPNA4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KPNA4. Alternate names: IPOA3, MGC12217, MGC26703, QIP1, SRP3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53742