Learn More
ABclonal Technology KPNA6 Rabbit mAb
SDP
Catalog No. 5000011316
Encompass_Preferred
Nucleocytoplasmic transport a signal- and energy-dependent process takes place through nuclear pore complexes embedded in the nuclear envelope The import of proteins containing a nuclear localization signal (NLS) requires the NLS import receptor a heterodimer of importin alpha and beta subunits also known as karyopherins Importin alpha binds the NLS-containing cargo in the cytoplasm and importin beta docks the complex at the cytoplasmic side of the nuclear pore complex In the presence of nucleoside triphosphates and the small GTP binding protein Ran the complex moves into the nuclear pore complex and the importin subunits dissociate Importin alpha enters the nucleoplasm with its passenger protein and importin beta remains at the pore The protein encoded by this gene is a member of the importin alpha family
Catalog No. 50-000-11316 Supplier ABclonal Technology Supplier No. A2482620UL
Nucleocytoplasmic transport a signal- and energy-dependent process takes place through nuclear pore complexes embedded in the nuclear envelope The import of proteins containing a nuclear localization signal (NLS) requires the NLS import receptor a heterodimer of importin alpha and beta subunits also known as karyopherins Importin alpha binds the NLS-containing cargo in the cytoplasm and importin beta docks the complex at the cytoplasmic side of the nuclear pore complex In the presence of nucleoside triphosphates and the small GTP binding protein Ran the complex moves into the nuclear pore complex and the importin subunits dissociate Importin alpha enters the nucleoplasm with its passenger protein and importin beta remains at the pore The protein encoded by this gene is a member of the importin alpha family