Learn More
Aviva Systems Biology Corp KPNB1 Antibody - N-terminal region (ARP61178_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61178_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KPNB1. It was validated on Western Blot. . Alternate names: IMB1, IPO1, IPOB, Impnb, MGC2155, MGC2156, MGC2157, NTF97ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-284