Learn More
Apexbio Technology LLC KPT-185, 10mg. Cas: 1333151-73-7 MFCD: MFCD25976761. CRM1 inhibitor,selective and irrversible
SDP
Supplier:  Apexbio Technology LLC B146210
Encompass_Preferred
KPT-185 is a selective and irreversible inhibitor of chromosome maintenance protein 1 (CRM1) with IC50 values of 100-500nM [1].CRM1 is an important nuclear protein export receptor. Blocking CRM1-mediated nuclear export can result in the suppression of tumors. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5240