Learn More
Apexbio Technology LLC KPT-185 1333151-73-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063658
Encompass_Preferred
KPT-185 (CAS 1333151-73-7) is an irreversible selective inhibitor of chromosome region maintenance 1 (CRM1) a nuclear export receptor essential for nuclear-cytoplasmic transport KPT-185 binds covalently at Cys528 of CRM1 thereby inhibiting CRM1-mediated nuclear export of tumor suppressor proteins Studies demonstrate that KPT-185 impairs proliferation and colony formation in acute myeloid leukemia (AML) cell lines exhibiting IC50 values ranging from 100 500 nM inducing G1 cell cycle arrest apoptosis and differentiation of AML blasts Additionally KPT-185 reduces growth and promotes apoptosis in pancreatic cancer models (Colo-357 HPAC BxPC-3) highlighting its potential in cancer research
Catalog No. 50-000-63658 Supplier Apexbio Technology LLC Supplier No. B14625.1
KPT-185 (CAS 1333151-73-7) is an irreversible selective inhibitor of chromosome region maintenance 1 (CRM1) a nuclear export receptor essential for nuclear-cytoplasmic transport KPT-185 binds covalently at Cys528 of CRM1 thereby inhibiting CRM1-mediated nuclear export of tumor suppressor proteins Studies demonstrate that KPT-185 impairs proliferation and colony formation in acute myeloid leukemia (AML) cell lines exhibiting IC50 values ranging from 100 500 nM inducing G1 cell cycle arrest apoptosis and differentiation of AML blasts Additionally KPT-185 reduces growth and promotes apoptosis in pancreatic cancer models (Colo-357 HPAC BxPC-3) highlighting its potential in cancer research