Learn More
Apexbio Technology LLC KPT-330 1393477-72-9 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061994
Encompass_Preferred
KPT-330 (CAS 1393477-72-9) is an orally bioavailable small-molecule inhibitor selectively targeting chromosome region maintenance 1 protein (CRM1) a nuclear export receptor involved in the active transport of tumor suppressors transcription factors and cell-cycle regulators Overexpression and deregulation of CRM1 are frequently associated with cancer progression KPT-330 inhibits CRM1 function promoting nuclear retention of regulatory proteins such as p21 and inducing apoptosis through activation of apoptotic signaling pathways (e g PAR-4 activation Bax upregulation PARP cleavage caspase-3 activation) In preclinical studies KPT-330 demonstrated antitumor activity against renal cell carcinoma non-small cell lung cancer and pancreatic cancer models
Catalog No. 50-000-61994 Supplier Apexbio Technology LLC Supplier No. B14645.1
KPT-330 (CAS 1393477-72-9) is an orally bioavailable small-molecule inhibitor selectively targeting chromosome region maintenance 1 protein (CRM1) a nuclear export receptor involved in the active transport of tumor suppressors transcription factors and cell-cycle regulators Overexpression and deregulation of CRM1 are frequently associated with cancer progression KPT-330 inhibits CRM1 function promoting nuclear retention of regulatory proteins such as p21 and inducing apoptosis through activation of apoptotic signaling pathways (e g PAR-4 activation Bax upregulation PARP cleavage caspase-3 activation) In preclinical studies KPT-330 demonstrated antitumor activity against renal cell carcinoma non-small cell lung cancer and pancreatic cancer models