Learn More
Medchemexpress LLC HY-123847 1mg Medchemexpress, KPT-6566 CAS:881487-77-0 Purity:>98%
SDP
Catalog No. 501969649
Encompass_Preferred
Medchemexpress, HY-123847 1mg KPT-6566 CAS:881487-77-0 KPT-6566, a potent prolyl isomerase PIN1 inhibitor, covalently binds to the catalytic site of PIN1, selectively inhibits and degrades PIN1. KPT-6566 shows an IC50 of 640 nM and a Ki of 625.2 nM for PIN1 PPIase domain. Anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9649 Supplier Medchemexpress LLC Supplier No. HY1238471MG
Medchemexpress, HY-123847 1mg KPT-6566 CAS:881487-77-0 KPT-6566, a potent prolyl isomerase PIN1 inhibitor, covalently binds to the catalytic site of PIN1, selectively inhibits and degrades PIN1. KPT-6566 shows an IC50 of 640 nM and a Ki of 625.2 nM for PIN1 PPIase domain. Anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.