Learn More
Medchemexpress LLC KPT-6566 | 881487-77-0 | 98.0% | 443.54 g/mol | C22H21NO5S2 | 25MG
SDP
Catalog No. 5000237486
Encompass_Preferred
KPT-6566 is a selective, covalent inhibitor of the prolyl isomerase PIN1 intended for laboratory research. It binds covalently to the catalytic site of PIN1, inhibiting enzyme activity and promoting PIN1 degradation; reported to induce DNA damage and produce antiproliferative effects in cancer cell models. Supplied as a solid with reported purity around 98.0% and commonly used in in vitro studies.
  • Selective covalent inhibitor of PIN1.
  • Promotes degradation of PIN1 and inhibits its enzymatic activity.
  • Induces DNA damage and elicits antiproliferative effects in cell models.
  • Supplied as a solid with typical purity of 98.0%.
  • Soluble in DMSO and provided in multiple milligram sizes suitable for in vitro work.
  • For research use only; not for human or diagnostic use.
Catalog No. 50-002-37486 Supplier Medchemexpress LLC Supplier No. HY12384725MG
KPT-6566 is a selective, covalent inhibitor of the prolyl isomerase PIN1 intended for laboratory research. It binds covalently to the catalytic site of PIN1, inhibiting enzyme activity and promoting PIN1 degradation; reported to induce DNA damage and produce antiproliferative effects in cancer cell models. Supplied as a solid with reported purity around 98.0% and commonly used in in vitro studies.
  • Selective covalent inhibitor of PIN1.
  • Promotes degradation of PIN1 and inhibits its enzymatic activity.
  • Induces DNA damage and elicits antiproliferative effects in cell models.
  • Supplied as a solid with typical purity of 98.0%.
  • Soluble in DMSO and provided in multiple milligram sizes suitable for in vitro work.
  • For research use only; not for human or diagnostic use.