Learn More
Selleck Chemical LLC KPT 9274 ( ATG-019) 2mg 1643913-93-2
SDP
Supplier:  Selleck Chemical LLC S84442MG
Encompass_Preferred
KPT 9274 ( ATG-019) is an orally bioavailable small molecule that is a non-competitive dual inhibitor of PAK4 and NAMPT. It shows an IC50 of ~120 nM for NAMPT in a cell-free enzymatic assay. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8232