Learn More
TARGETMOL CHEMICALS INC KPT276 25MG
SDP
Catalog No. 502576608
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. KPT276 (KPT 276) is an orally bioavailable selective inhibitor of nuclear export (SINE) that irreversibly bind to CRM1 and block the function of CRM1. purity: 98%
Catalog No. 50-257-6608 Supplier TARGETMOL CHEMICALS INC Supplier No. T607425MG
Also available in 5 mg 10 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. KPT276 (KPT 276) is an orally bioavailable selective inhibitor of nuclear export (SINE) that irreversibly bind to CRM1 and block the function of CRM1. purity: 98%