Learn More
TARGETMOL CHEMICALS INC KPT9274 5MG
SDP
Catalog No. 502579900
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. KPT9274 (PAK4-IN-1) is a non-competitive dual inhibitor of PAK4 and NAMPT(IC50= ~120 nM). It is an orally bioavailable small molecule. purity: 99%
Catalog No. 50-257-9900 Supplier TARGETMOL CHEMICALS INC Supplier No. T43545MG
Add to Cart
Add to Cart
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. KPT9274 (PAK4-IN-1) is a non-competitive dual inhibitor of PAK4 and NAMPT(IC50= ~120 nM). It is an orally bioavailable small molecule. purity: 99%