Learn More
Krackeler Scientific GC and GC/MS Packed Column Inlet Accessories: Disposable Liner
Specifications
Specifications
| For Use With (Equipment) | GC 7890/6890/6850 Inlet |
| Material | Glass |
| Volume (Metric) | 170 μL |
| Product Type | Liner |