Learn More
Krackeler Scientific Glass Flat Bottom Inserts for GC
SDP
Catalog No. 51813377N
Change view
Click to view available options
Volume (Metric):
0.25 mL
Catalog No. Volume (Metric)
51813377N 0.25 mL
1 options
Catalog No. 51813377N Supplier Krackeler Scientific Supplier No. 9251813377EA
Add to Cart
Add to Cart
Microvolume inserts for wide opening vials
  • Glass
  • Volume: 250μL
Specifications
Material Glass
Volume (Metric) 0.25 mL
For Use With (Application) GC, used to transform vials from full-capacity to limited volume with one simple step, compatible with wide opening screw top, crimp top and snap top vials
No. per Case 500
Base Style Flat
Quantity 500 Cs.