Learn More
Krackeler Scientific Screw Caps without Septa
SDP
Catalog No. 93010718N
Change view
Click to view available options
Includes:
Safety Release
Catalog No. Includes
93010718N Safety Release
1 options
Catalog No. 93010718N Supplier Krackeler Scientific Supplier No. 9293010718EA
Add to Cart
Add to Cart
For crimp top vials
  • 20mm headspace
  • Silver aluminum
  • Also available with safety feature
Specifications
Color Silver
Material Aluminum
Includes Safety Release
Quantity 100 Pk.
Closure Size 20 mm
Product Type Crimp Cap