Learn More
Krackeler Scientific Tubing Cutter
SDP
Catalog No. 51818836N
Change view
Click to view available options
Ceramic capillary tubing cutter
Catalog No.
51818836N Ceramic capillary tubing cutter
1 options
Catalog No. 51818836N Supplier Krackeler Scientific Supplier No. 9251818836EA
Add to Cart
Add to Cart
Enables clean and easy cuts
  • Capillary column cutter with ceramic wafer
  • Cuts fused silica, glass and aluminum-clad capillary columns
Specifications
For Use With Used silica, glass and aluminum-clad capillary columns
For Use With (Application) Capillary column cutter
Product Type Tubing Cutter