Learn More
Krackeler Scientific UV-Vis Cells and Cell Accessories
SDP
Catalog No. 50613387N
Change view
Click to view available options
Type:
Macro Cell
Catalog No. Type
50613387N Macro Cell
1 options
Catalog No. 50613387N Supplier Krackeler Scientific Supplier No. 9250613387EA
Specifications
Type Macro Cell
For Use With (Equipment) 8453 UV-vis spectrophotometer
Includes PTFE Lid
Material Quartz
Pathlength 10 mm
Shape Rectangular
Volume (Metric) 3.5 mL