Learn More
MSC Kranzle Pressure Washer, 5 GPM, Hot Water, Electric, 2400 PSI
Catalog No. 502836936
Encompass
This hot water electric pressure washer is designed for industrial and commercial cleaning applications such as factories, food processing plants, vehicle wash bays, farms, and rental stores.
  • Advanced pump design for multi-hour bypass and run-dry capability.
  • Industrial turbo nozzle doubles cleaning efficiency.
  • High-pressure soap design delivers 20 percent more cleaning power.
  • Auto on/off control at gun jet with 30-second time delay.
  • Digital temperature control for precise hot water application.
  • Multiple safety features, including optical flame monitoring.
  • Downdraft coil-burner system for maximum efficiency.
  • 65-foot wire braided hose on a hose reel with safety couplers.
  • TEFC 1600 RPM electric 220V, 25A, three-phase motor.
  • Uses diesel fuel for water heating.
Catalog No. 50-283-6936 Supplier MSC Supplier No. 64996770
Add to Cart
Add to Cart
Request a Quote
This hot water electric pressure washer is designed for industrial and commercial cleaning applications such as factories, food processing plants, vehicle wash bays, farms, and rental stores.
  • Advanced pump design for multi-hour bypass and run-dry capability.
  • Industrial turbo nozzle doubles cleaning efficiency.
  • High-pressure soap design delivers 20 percent more cleaning power.
  • Auto on/off control at gun jet with 30-second time delay.
  • Digital temperature control for precise hot water application.
  • Multiple safety features, including optical flame monitoring.
  • Downdraft coil-burner system for maximum efficiency.
  • 65-foot wire braided hose on a hose reel with safety couplers.
  • TEFC 1600 RPM electric 220V, 25A, three-phase motor.
  • Uses diesel fuel for water heating.