Learn More
Aviva Systems Biology Corp Kras Antibody - N-terminal region (ARP55409_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55409_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Kras. It was validated on Western Blot. . Alternate names: Kras2, c-Ki-ras, p21ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-031