Learn More
AURORA BIOLABS LLC Kras G12C, GppNHp loaded, 100 μg
Catalog No. NC3787930
Encompass
Kras is a member of the small G protein RAS family, which are a class of GTPases involved in cell signaling transduction. As an oncogene mutated in several different cancers, the G12C substitution mutation favors the activated, GTP-bound state of the protein.
Catalog No. NC3787930 Supplier AURORA BIOLABS LLC Supplier No. 57274122G GP100
Kras is a member of the small G protein RAS family, which are a class of GTPases involved in cell signaling transduction. As an oncogene mutated in several different cancers, the G12C substitution mutation favors the activated, GTP-bound state of the protein.