Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC KRAS G12C inhibitor 14 | 2349393-95-7 | 99.0% | 484.88 | C24H19ClF2N4O3 | 10MM 1ML
SDP

Catalog No. 5000235600
Encompass_Preferred

KRAS G12C inhibitor 14 (Compound 17) is a potent small-molecule inhibitor selective for the KRAS G12C mutant (reported IC50 = 18 nM). It is supplied as a solid and as a 10 mM solution in DMSO for research use and is suitable for in vitro and in vivo studies of KRAS-driven signaling.

  • Potency: IC50 = 18 nM against KRAS G12C.
  • Molecular formula: C24H19ClF2N4O3; molecular weight: 484.88 g/mol.
  • Purity: 99.0%.
  • Available formats: solid (1 mg-500 mg) and 10 mM, 1 mL solution in DMSO.
  • Solubility: DMSO 100 mg/mL; in vivo formulation yields ≥9 mg/mL in 10% DMSO/40% PEG300/5% Tween-80/45% saline.
  • Storage: solid at -20°C; stock solution -80°C (6 months) or -20°C (1 month); protect from light and avoid repeated freeze-thaw.

Catalog No. 50-002-35600 Supplier Medchemexpress LLC Supplier No. HY12587210MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KRAS G12C inhibitor 14 (Compound 17) is a potent small-molecule inhibitor selective for the KRAS G12C mutant (reported IC50 = 18 nM). It is supplied as a solid and as a 10 mM solution in DMSO for research use and is suitable for in vitro and in vivo studies of KRAS-driven signaling.

  • Potency: IC50 = 18 nM against KRAS G12C.
  • Molecular formula: C24H19ClF2N4O3; molecular weight: 484.88 g/mol.
  • Purity: 99.0%.
  • Available formats: solid (1 mg-500 mg) and 10 mM, 1 mL solution in DMSO.
  • Solubility: DMSO 100 mg/mL; in vivo formulation yields ≥9 mg/mL in 10% DMSO/40% PEG300/5% Tween-80/45% saline.
  • Storage: solid at -20°C; stock solution -80°C (6 months) or -20°C (1 month); protect from light and avoid repeated freeze-thaw.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.