Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC KRAS G12C inhibitor 14 | 2349393-95-7 | 99.0% | 484.88 g/mol | C24H19ClF2N4O3 | 1MG
SDP

Catalog No. 5000236117
Encompass_Preferred

KRas G12C inhibitor 14 is a small-molecule covalent inhibitor selective for KRAS G12C designed for research applications. It demonstrates potent biochemical activity and includes manufacturer guidance for solubility, storage, and formulation for in vitro and in vivo work.

  • Potent KRAS G12C inhibition (IC50 = 18 nM).
  • Chemical formula C24H19ClF2N4O3.
  • Molecular weight 484.88 g/mol.
  • Purity 99.0%.
  • CAS number 2349393-95-7.
  • Available sizes: 1 mg, 5 mg, 10 mg.
  • DMSO solubility 100 mg/mL; in vivo formulation guidance provided.
  • Storage: -20°C, protect from light, store under nitrogen; in solvent: -80°C (6 months) or -20°C (1 month).
  • Target: Ras; relevant to MAPK/ERK pathway.
  • For research use only; not for clinical or human use.

Catalog No. 50-002-36117 Supplier Medchemexpress LLC Supplier No. HY1258721MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KRas G12C inhibitor 14 is a small-molecule covalent inhibitor selective for KRAS G12C designed for research applications. It demonstrates potent biochemical activity and includes manufacturer guidance for solubility, storage, and formulation for in vitro and in vivo work.

  • Potent KRAS G12C inhibition (IC50 = 18 nM).
  • Chemical formula C24H19ClF2N4O3.
  • Molecular weight 484.88 g/mol.
  • Purity 99.0%.
  • CAS number 2349393-95-7.
  • Available sizes: 1 mg, 5 mg, 10 mg.
  • DMSO solubility 100 mg/mL; in vivo formulation guidance provided.
  • Storage: -20°C, protect from light, store under nitrogen; in solvent: -80°C (6 months) or -20°C (1 month).
  • Target: Ras; relevant to MAPK/ERK pathway.
  • For research use only; not for clinical or human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.