Learn More
Medchemexpress LLC KRAS G12C inhibitor 15 | 2349393-21-9 | 98.5% | 498.91 | C25H21ClF2N4O3 | 10MG
SDP
Catalog No. 5000237827
Encompass_Preferred
KRAS G12C inhibitor 15 is a potent small-molecule inhibitor of the oncogenic KRAS G12C mutant, reported from patent WO2019110751A1 (compound 22). It is supplied as a high-purity solid and as a DMSO solution for research use, and biochemical assays report an IC50 of 5 nM for KRAS G12C.
  • Potent KRAS G12C inhibition (IC50 = 5 nM).
  • High purity, 98.5%.
  • Molecular weight 498.91 and formula C25H21ClF2N4O3.
  • Available in small solid quantities and as a 10 mM solution in DMSO.
  • Suitable for biochemical and cell-based research applications.
Catalog No. 50-002-37827 Supplier Medchemexpress LLC Supplier No. HY12587310MG
KRAS G12C inhibitor 15 is a potent small-molecule inhibitor of the oncogenic KRAS G12C mutant, reported from patent WO2019110751A1 (compound 22). It is supplied as a high-purity solid and as a DMSO solution for research use, and biochemical assays report an IC50 of 5 nM for KRAS G12C.
  • Potent KRAS G12C inhibition (IC50 = 5 nM).
  • High purity, 98.5%.
  • Molecular weight 498.91 and formula C25H21ClF2N4O3.
  • Available in small solid quantities and as a 10 mM solution in DMSO.
  • Suitable for biochemical and cell-based research applications.