Learn More
Medchemexpress LLC KRAS G12C inhibitor 15 | 2349393-21-9 | 98.5% | 498.91 | C25H21ClF2N4O3 | 1MG
SDP
Catalog No. 5000236118
Encompass_Preferred
KRAS G12C inhibitor 15 is a small-molecule inhibitor targeting the KRAS G12C mutant, reported to have an IC50 of 5 nM. The compound is provided as a solid with high purity for research use and includes supporting documentation for handling and analysis.
Catalog No. 50-002-36118 Supplier Medchemexpress LLC Supplier No. HY1258731MG
KRAS G12C inhibitor 15 is a small-molecule inhibitor targeting the KRAS G12C mutant, reported to have an IC50 of 5 nM. The compound is provided as a solid with high purity for research use and includes supporting documentation for handling and analysis.