Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Kras G12c inhibitor 15 | 2349393-21-9 | 98.5% | 498.91 | C25H21ClF2N4O3 | 25MG
SDP

Catalog No. 5000239092
Encompass_Preferred

KRAS G12C inhibitor 15 is a research-grade small-molecule inhibitor of the oncogenic KRAS G12C mutant. It was reported in patent WO2019110751A1 (compound 22) and shows potent biochemical activity; supplied as a solid for laboratory research only - consult the SDS and COA for handling and storage.

  • Potent KRAS G12C inhibition (IC50 ≈ 5 nM).
  • High purity (98.5%).
  • Molecular formula C25H21ClF2N4O3.
  • Molecular weight 498.91 g/mol.
  • Appearance solid, white to yellow.
  • Storage: protect from light and store under nitrogen; in solvent -80°C (6 months) or -20°C (1 month).
  • Available as solid and as 10 mM solutions in DMSO; common pack sizes up to 100 mg.

Catalog No. 50-002-39092 Supplier Medchemexpress LLC Supplier No. HY12587325MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KRAS G12C inhibitor 15 is a research-grade small-molecule inhibitor of the oncogenic KRAS G12C mutant. It was reported in patent WO2019110751A1 (compound 22) and shows potent biochemical activity; supplied as a solid for laboratory research only - consult the SDS and COA for handling and storage.

  • Potent KRAS G12C inhibition (IC50 ≈ 5 nM).
  • High purity (98.5%).
  • Molecular formula C25H21ClF2N4O3.
  • Molecular weight 498.91 g/mol.
  • Appearance solid, white to yellow.
  • Storage: protect from light and store under nitrogen; in solvent -80°C (6 months) or -20°C (1 month).
  • Available as solid and as 10 mM solutions in DMSO; common pack sizes up to 100 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.