Learn More
Medchemexpress LLC KRAS G12C inhibitor 15 | 2349393-21-9 | 98.5% | 498.91 g/mol | C25H21ClF2N4O3 | 100MG
SDP
Catalog No. 5000238238
Encompass_Preferred
KRAS G12C inhibitor 15 is a research-grade small-molecule inhibitor selective for the KRAS G12C mutant, derived from patent WO2019110751A1 (compound 22). It is reported to inhibit KRAS G12C with an IC50 of 5 nM and is supplied as a solid for laboratory research use only.
  • Potent KRAS G12C inhibition (IC50 5 nM).
  • High purity (~98.5%).
  • Molecular weight 498.91 g/mol; formula C25H21ClF2N4O3.
  • White to yellow solid suitable for biochemical and cellular assays.
  • Supplied as 100 MG for research use only.
  • Derived from patent WO2019110751A1 (compound 22).
Catalog No. 50-002-38238 Supplier Medchemexpress LLC Supplier No. HY125873100MG
Add to Cart
Add to Cart
Request a Quote
KRAS G12C inhibitor 15 is a research-grade small-molecule inhibitor selective for the KRAS G12C mutant, derived from patent WO2019110751A1 (compound 22). It is reported to inhibit KRAS G12C with an IC50 of 5 nM and is supplied as a solid for laboratory research use only.
  • Potent KRAS G12C inhibition (IC50 5 nM).
  • High purity (~98.5%).
  • Molecular weight 498.91 g/mol; formula C25H21ClF2N4O3.
  • White to yellow solid suitable for biochemical and cellular assays.
  • Supplied as 100 MG for research use only.
  • Derived from patent WO2019110751A1 (compound 22).